Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC4H2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113394
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113394 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113394 Supplier Invitrogen™ Supplier No. PA5113394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZC4H2 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

May play a role in neuronal development and in neuromuscular junction formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC4H2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZC4H2
Gene Accession No. Q68FG0, Q9NQZ6
Gene Alias Gm372; HCA127; hepatocellular carcinoma-associated antigen 127; Hepatocellular carcinoma-associated antigen 127 homolog; KIAA1166; mKIAA1166; WRWF; WWS; Zc4h2; Zinc finger C4H2 domain-containing protein; zinc finger C4H2-type containing; zinc finger, C4H2 domain containing
Gene Symbols ZC4H2
Host Species Rabbit
Immunogen Recombinant Human Zinc finger C4H2 domain-containing protein (1-224AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245522, 55906
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.