Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZCCHC10 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306032IR
Description
ZCCHC10 Polyclonal specifically detects ZCCHC10 in Human samples. It is validated for Western Blot.Specifications
ZCCHC10 | |
Polyclonal | |
Western Blot | |
FLJ20094, zinc finger CCHC domain-containing protein 10, zinc finger, CCHC domain containing 10 | |
Produced in rabbits immunized with E. coli-derived Human ZCCHC10 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
54819 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction