Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZCCHC10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558602
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558602 Supplier Invitrogen™ Supplier No. PA558602
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WTYECTGKRK YLHRPSRTAE LKKALKEKEN RLLLQQSIGE TNVERKAKKK RSKSVTS Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.

A member of the family of Zcchc (zinc-finger, CCHC domain-containing protein) proteins. Zcchc contains RNA-interacting motifs, and has been implicated in signalling pathways involved in cytokine expression (Hagan JP, et al. (2009) Nat Struct Mol Biol.16(10):1021-5. Titulaer MK, et al. (2008) BMC Bioinformatics.9:133. Li H, et al.(2009) Acta Biochim Biophys Sin (Shanghai).41(7):535-44).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCCHC10
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZCCHC10
Gene Accession No. Q8TBK6
Gene Alias 2410141K03Rik; AA675321; D11Ertd416e; FLJ20094; Zcchc10; Zinc finger CCHC domain-containing protein 10; zinc finger CCHC-type containing 10; zinc finger, CCHC domain containing 10
Gene Symbols ZCCHC10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZCCHC10. Recombinant protein control fragment (Product #RP-96908).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54819
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.