Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZCCHC10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549677
Description
The antibody detects endogenous level of total Zinc finger CCHC domain-containing protein.
A member of the family of Zcchc (zinc-finger, CCHC domain-containing protein) proteins. Zcchc contains RNA-interacting motifs, and has been implicated in signalling pathways involved in cytokine expression (Hagan JP, et al. (2009) Nat Struct Mol Biol.16(10):1021-5. Titulaer MK, et al. (2008) BMC Bioinformatics.9:133. Li H, et al.(2009) Acta Biochim Biophys Sin (Shanghai).41(7):535-44).
Specifications
| ZCCHC10 | |
| Polyclonal | |
| Unconjugated | |
| ZCCHC10 | |
| 2410141K03Rik; AA675321; D11Ertd416e; FLJ20094; Zcchc10; Zinc finger CCHC domain-containing protein 10; zinc finger CCHC-type containing 10; zinc finger, CCHC domain containing 10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54819 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8TBK6 | |
| ZCCHC10 | |
| Peptide sequence around aa.10~14 (A-R-R-Q-A) derived from Human Zinc finger CCHC domain-containing protein 10. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction