Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZCCHC11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544931 Supplier Invitrogen™ Supplier No. PA544931
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GSAQQQGDQS IRTRQSSECS ESPSYSPQPQ PFPQNSSQSA AITQPSSQPG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 92%; Rabbit: 92%; Rat: 100%.

WDR77 is a component of the 20S PRMT5-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins. This modification targets Sm proteins to the survival of motor neurons (SMN) complex for assembly into small nuclear ribonucleoprotein core particles (Friesen et al., 2002).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCCHC11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TUT4
Gene Accession No. Q5TAX3
Gene Alias 6030404K05Rik; 9230115F04Rik; Kiaa0191; mKIAA0191; PAP associated domain containing 3; PAPD3; PPAPD3; RP23-406B13.1; terminal uridylyl transferase 4; terminal uridylyltransferase 4; terminal uridylyltransferase 4-like; TUT4; TUTase 4; ZCCHC11; Zinc finger CCHC domain-containing protein 11; zinc finger CCHC-type containing 11; zinc finger, CCHC domain containing 11
Gene Symbols TUT4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZCCHC11.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23318
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.