Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZCCHC11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555604
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555604 Supplier Invitrogen™ Supplier No. PA555604
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VARSLNSQLV YEYVVERFRA AYRYFACPQT KGGNKSTVDF KKREKGKISN KKPVKSNNMA TNGCILLGET TEKINAEREQ PVQCDEMDC Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 80%.

WDR77 is a component of the 20S PRMT5-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins. This modification targets Sm proteins to the survival of motor neurons (SMN) complex for assembly into small nuclear ribonucleoprotein core particles (Friesen et al., 2002).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCCHC11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TUT4
Gene Accession No. Q5TAX3
Gene Alias 6030404K05Rik; 9230115F04Rik; Kiaa0191; mKIAA0191; PAP associated domain containing 3; PAPD3; PPAPD3; RP23-406B13.1; terminal uridylyl transferase 4; terminal uridylyltransferase 4; terminal uridylyltransferase 4-like; TUT4; TUTase 4; ZCCHC11; Zinc finger CCHC domain-containing protein 11; zinc finger CCHC-type containing 11; zinc finger, CCHC domain containing 11
Gene Symbols TUT4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZCCHC11. Recombinant protein control fragment (Product #RP-94218).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23318
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.