Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZCCHC11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555739
Description
Immunogen sequence: CNCIDLSKSP DPDKSTGTDC RSNLETESSH QSVCTDTSAT SCNCKATEDA SDLNDDDNLP TQELYYVFDK FILTSGK Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 75%.
WDR77 is a component of the 20S PRMT5-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins. This modification targets Sm proteins to the survival of motor neurons (SMN) complex for assembly into small nuclear ribonucleoprotein core particles (Friesen et al., 2002).
Specifications
| ZCCHC11 | |
| Polyclonal | |
| Unconjugated | |
| TUT4 | |
| 6030404K05Rik; 9230115F04Rik; Kiaa0191; mKIAA0191; PAP associated domain containing 3; PAPD3; PPAPD3; RP23-406B13.1; terminal uridylyl transferase 4; terminal uridylyltransferase 4; terminal uridylyltransferase 4-like; TUT4; TUTase 4; ZCCHC11; Zinc finger CCHC domain-containing protein 11; zinc finger CCHC-type containing 11; zinc finger, CCHC domain containing 11 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23318 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5TAX3 | |
| TUT4 | |
| Recombinant protein corresponding to Human ZCCHC11. Recombinant protein control fragment (Product #RP-94219). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction