Learn More
ZCCHC11 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302635AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
ZCCHC11 is an RNA uridyltransferase that uses UTP to add uridines to the 3-prime end of substrate RNA molecules .Specifications
ZCCHC11 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q5TAX3 | |
ZCCHC11 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ZCCHC11 | |
ZCCHC11; KIAA0191; zinc finger, CCHC domain containing 11; PAPD3; PAP associated domain containing 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23318 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.