Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZCCHC11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 50554585 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-554-585 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-554-585 Supplier Proteintech Group Inc Supplier No. 189801AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is specifically to the 185 kDa ZCCHCC11 protein.

WDR77 is a component of the 20S PRMT5-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins. This modification targets Sm proteins to the survival of motor neurons (SMN) complex for assembly into small nuclear ribonucleoprotein core particles (Friesen et al., 2002).

Specifications

Antigen ZCCHC11
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZCCHC11
Gene Accession No. B2RX14, Q5TAX3
Gene Alias KIAA0191, PAPD3, Terminal uridylyltransferase 4, TUT4, TUTase 4, ZCCHC11
Gene Symbols Zcchc11
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230594, 23318
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.