Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZCCHC12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114001
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114001 Supplier Invitrogen™ Supplier No. PA5114001
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKASLYVIRL EVQLQNAIQA GIIAEKDANR TRLQQLLLGG ELSRDLRLRL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Transcriptional coactivator in the bone morphogenetic protein (BMP)-signaling pathway. It positively modulates BMP signaling by interacting with SMAD1 and associating with CBP in the transcription complex. It contributes to the BMP-induced enhancement of cholinergic-neuron-specific gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCCHC12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZCCHC12
Gene Accession No. Q6PEW1
Gene Alias paraneoplastic Ma antigen family member 7A; PNMA7A; SIZN; SIZN1; Smad-interacting zinc finger protein 1; ZCCHC12; Zinc finger CCHC domain-containing protein 12; zinc finger CCHC-type containing 12; zinc finger, CCHC domain containing 12
Gene Symbols ZCCHC12
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZCCHC12.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170261
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.