Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZCCHC13 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | ZCCHC13 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZCCHC13 Polyclonal specifically detects ZCCHC13 in Human samples. It is validated for Western Blot.Specifications
| ZCCHC13 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 4930513O09RIK, Cnbp2, zinc finger CCHC domain-containing protein 13, zinc finger, CCHC domain containing 13, ZNF9L | |
| ZCCHC13 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q8WW36 | |
| 389874 | |
| Synthetic peptides corresponding to ZCCHC13(zinc finger, CCHC domain containing 13) The peptide sequence was selected from the middle region of ZCCHC13. Peptide sequence GKLGHIQKDCAQVKCYRCGEIGHVAINCSKARPGQLLPLRQIPTSSQGMS. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title