Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZCCHC13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551918
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551918 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551918 Supplier Invitrogen™ Supplier No. PA551918
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGGRRGGGHG RGSQCGSTTL SYTCYCCGES GRNAKNCVLL GNICYNCGRS GHIAKDCKDP KRERRQHCYT CGRLGHLARD CDRQKEQKCY SCGKLGHIQK DCAQVKCYRC Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 65%.

This gene appears to represent an intronless retrocopy of a related multi-exon gene located on chromosome 3. However, the CDS of this intronless gene remains relatively intact, it is conserved in other mammalian species, it is known to be transcribed, and it is therefore thought to encode a functional protein. The encoded protein contains six CCHC-type zinc fingers, and is thus thought to function as a transcription factor. [provided by RefSeq, May 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCCHC13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZCCHC13
Gene Accession No. Q8WW36
Gene Alias CNBP2; ZCCHC13; Zinc finger CCHC domain-containing protein 13; zinc finger CCHC-type containing 13; zinc finger, CCHC domain containing 13; ZNF9L
Gene Symbols ZCCHC13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZCCHC13. Recombinant protein control fragment (Product #RP-92270).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389874
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.