Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZCCHC14 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171852IR
Description
ZCCHC14 Polyclonal antibody specifically detects ZCCHC14 in Human samples. It is validated for Western BlotSpecifications
ZCCHC14 | |
Polyclonal | |
Western Blot | |
BDG29, BDG-29, KIAA0579, MGC126527, MGC14139, zinc finger CCHC domain-containing protein 14, zinc finger, CCHC domain containing 14 | |
The immunogen for this product maps to a region between residue 899 and 949 of human Zinc Finger, CCHC Domain Containing 14 using the numbering given in entry NP_055959.1 (GeneID 23174). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23174 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction