Learn More
ZCCHC17 Mouse anti-Human, Clone: OTI9A7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809551

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. ZCCHC17 (zinc finger, CCHC domain containing 17), also known as PS1D (putative S1 RNA-binding domain protein), Pnn (Pinin)-interacting nucleolar protein or pNO40, is a 241 amino acid protein that associates with both Pinin and the 60S ribosomal subunit. Localizing to nucleolus, ZCCHC17 is ubiquitously expressed and has been suggested to play a role in ribosome maturation and biogenesis. ZCCHC17 contains one CCHC-type zinc finger, a S1 motif domain and exists as two alternatively spliced isoforms that map to human chromosome 1p35.2. Human chromosome 1 spans 260 million base pairs, contains over 3,000 genes, comprises nearly 8% of the human genome and houses a large number of disease-associated genes, including those that are involved in Stickler syndrome, Parkinson′s disease, Gaucher disease and Usher syndrome.Specifications
ZCCHC17 | |
Monoclonal | |
Unconjugated | |
ZCCHC17 | |
HSPC251, PS1D, pNO40 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51538 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI9A7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NP64 | |
ZCCHC17 | |
Full length human recombit protein of human ZCCHC17 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.