Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZCCHC17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575895 Supplier Invitrogen™ Supplier No. PA575895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

ZCCHC17 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cytosolic large ribosomal subunit; nucleolus; nucleic acid binding; zinc ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCCHC17
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ZCCHC17
Gene Accession No. Q9ESX4, Q9NP64
Gene Alias 2810055E05Rik; hpNO40s; HSPC243; HSPC251; LDC4; nucleolar protein 40; nucleolar protein of 40 kDa; Pnn-interacting nucleolar protein; pNO40; PS1D; PS1D protein; putative S1 RNA binding domain protein; putative S1 RNA-binding domain protein; RGD1565267; Zcchc17; Zinc finger CCHC domain-containing protein 17; zinc finger CCHC-type containing 17; zinc finger, CCHC domain containing 17
Gene Symbols ZCCHC17
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human ZCCHC17.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 500555, 51538, 619605
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.