Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZCCHC17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584169 Supplier Invitrogen™ Supplier No. PA584169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNSGRPETME NLPALYTIFQ GEVAMVTDYG AFIKIPGCRK QGLVHRTHMS SCRVDKPSEI VDVGDKVWVK LIGREMK Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

ZCCHC17 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cytosolic large ribosomal subunit; nucleolus; nucleic acid binding; zinc ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCCHC17
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZCCHC17
Gene Accession No. Q9NP64
Gene Alias 2810055E05Rik; hpNO40s; HSPC243; HSPC251; LDC4; nucleolar protein 40; nucleolar protein of 40 kDa; Pnn-interacting nucleolar protein; pNO40; PS1D; PS1D protein; putative S1 RNA binding domain protein; putative S1 RNA-binding domain protein; RGD1565267; Zcchc17; Zinc finger CCHC domain-containing protein 17; zinc finger CCHC-type containing 17; zinc finger, CCHC domain containing 17
Gene Symbols ZCCHC17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZCCHC17. Recombinant protein control fragment (Product #RP-106491).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51538
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.