Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZCCHC17 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548118
Description

Specifications
ZCCHC17 | |
Polyclonal | |
Unconjugated | |
ZCCHC17 | |
Nucleolar protein of 40 kDa, pNO40, Pnn-interacting nucleolar protein, Putative S1 RNA-binding domain protein, PS1D protein, Zinc finger CCHC domain-containing protein 17, ZCCHC17, PS1D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51538, 619605 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q9NP64, Q9ESX4 | |
ZCCHC17 | |
KLH conjugated synthetic peptide between 159-187 amino acids from the Central region of human ZCCHC17. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction