Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZCCHC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587672
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587672 Supplier Invitrogen™ Supplier No. PA587672
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVTQGKEETR RFYACSACRD RKDCNFFQWE DEKLSGARLA AREAHNRRCQ PPLSRTQCVE RYLKFIELPL TQRKFCQTCQ QLLLPDDWGQ HSEHQVLGNV SITQLRRPSQ LLYPLENKKT NAQYLFADRS CQFLVDLLSA LGFRRVLCVG TPRLHELIKL TASGDKKSNI K; Positive Samples: Mouse kidney.

The ZCCHC4 gene encodes zinc finger CCHC domain-containing protein 4, a member of the zinc finger CCHC-type (ZCCHC) superfamily characterized by a consensus sequence C-X2-C-X4-H-X4-C. This gene plays a vital role in binding single-stranded nucleic acids, particularly RNAs, and is involved in multiple cellular processes. Notably, ZCCHC4 has been implicated in cancer, with aberrantly high expression linked to poor prognosis and chemoresistance in various cancers, including hepatocellular carcinoma, pancreatic cancer, colon cancer, and esophageal cancer. The protein exhibits RNA-modifying capabilities, influencing cell growth and survival. Recent studies have identified the structural mechanisms through which ZCCHC4 interacts with RNA substrates, offering insights into its role in regulating cancer progression and therapeutic resistance. In esophageal cancer, downregulation of ZCCHC4 may enhance sensitivity to chemotherapeutic agents like cisplatin, indicating its potential as a target for cancer treatment strategies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCCHC4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZCCHC4
Gene Accession No. Q8BKW4
Gene Alias 4930449I23Rik; DHHC domain-containing zinc finger protein; HSPC052; rRNA N6-adenosine-methyltransferase ZCCHC4; Zcchc4; ZGRF4; zinc finger CCHC domain-containing protein 4; zinc finger CCHC-type containing 4; zinc finger, CCHC domain containing 4; zinc finger, GRF-type containing 4
Gene Symbols ZCCHC4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 50-220 of human ZCCHC4 (NP_0792122).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78796
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.