Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZCCHC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554242
Description
Immunogen sequence: SNRWPQENKE TQKEMKNKNR NWEKHRKADR HREVDEDFPR GPKTYSSPGS FKTQKPSKPF HRSSHYHTSR EDKSPKEGKR GKQKKKERCW EDDDNDNLFL IKQ Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 53%.
The function of this protein remains unknown.
Specifications
| ZCCHC7 | |
| Polyclonal | |
| Unconjugated | |
| ZCCHC7 | |
| 4930572I07Rik; AIR1; D4Wsu132e; HSPC086; TRAMP-like complex RNA-binding factor ZCCHC7; ZCCHC7; Zinc finger CCHC domain-containing protein 7; zinc finger CCHC-type containing 7; zinc finger, CCHC domain containing 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84186 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N3Z6 | |
| ZCCHC7 | |
| Recombinant protein corresponding to Human ZCCHC7. Recombinant protein control fragment (Product #RP-92994). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction