Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZCCHC8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570266
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570266 Supplier Invitrogen™ Supplier No. PA570266
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.

ZCCHC8 may be involved in pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCCHC8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Zcchc8
Gene Accession No. 0
Gene Alias 5730565F05Rik; DKFZp434E2220; TRAMP-like complex RNA-binding factor ZCCHC8; Zcchc8; Zinc finger CCHC domain-containing protein 8; zinc finger CCHC-type containing 8; zinc finger, CCHC domain containing 8
Gene Symbols Zcchc8
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of rat ZCCHC8.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288661
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.