Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZCCHC8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583372
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583372 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583372 Supplier Invitrogen™ Supplier No. PA583372
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QYHQEIEEFV SNLVKRFEEQ QKNDVEKTSF NLLPQPSSIV LEEDHKVEES CAIKNNKEAF SVVGSVLYFT NFCLDKLGQP LLNENPQLSE GWEIPKYH Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 94%.

ZCCHC8 may be involved in pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCCHC8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Zcchc8
Gene Accession No. Q6NZY4, Q9CYA6
Gene Alias 5730565F05Rik; DKFZp434E2220; TRAMP-like complex RNA-binding factor ZCCHC8; Zcchc8; Zinc finger CCHC domain-containing protein 8; zinc finger CCHC-type containing 8; zinc finger, CCHC domain containing 8
Gene Symbols Zcchc8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZCCHC8. Recombinant protein control fragment (Product #RP-97631).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288661, 55596, 70650
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.