Learn More
ZCCHC8 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | ZCCHC8 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1948
![]() |
Bethyl Laboratories, Inc
IHC00353T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1947
![]() |
Bethyl Laboratories, Inc
IHC00353 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, Chimpanzee and Northern white-cheeked gibbon
ZCCHC8 may be involved in pre-mRNA splicing.Specifications
ZCCHC8 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
DKFZp434E2220, TRAMP-like complex RNA-binding factor ZCCHC8, zinc finger, CCHC domain containing 8 | |
ZCCHC8 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q6NZY4 | |
55596 | |
Between 1 and 50 | |
Primary | |
4° C | |
ZCCHC8 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.