Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZCRB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520772
Description
A suggested positive control is Raji cell lysate. PA5-20772 can be used with blocking peptide PEP-0886.
ZCRB1 is a nuclear protein first identified in a differential display screen involving morphine-dependence-related genes. Its expression is increased following morphine treatment and has been observed to be elevated in HepG2 cells. ZCRB1 contains a CCHC-type zing finger RNA-binding motif and can interact with the DBA-binding domain of the stem cell regulator C/EBP transcription factors. ZCRB1 was identified as one of the protein components of U11/U12 snRNPs, which are components of U12-type spliceosome and function as a molecular bridge connecting both ends of the intron, suggesting ZCRB1 may play a key role in U12-type splicing.
Specifications
ZCRB1 | |
Polyclonal | |
Unconjugated | |
Zcrb1 | |
2700088M22Rik; MADP1; Madp-1; MADP-1 protein; MADP-1 protein-like; MGC26805; RBM36; RGD1309851; SNRNP31; U11/U12 small nuclear ribonucleoprotein 31 kDa protein; U11/U12 snRNP 31 kDa protein; U11/U12 snRNP 31K; U11/U12-31K; ZCCHC19; Zcrb1; zinc finger CCHC-type and RNA binding motif 1; zinc finger CCHC-type and RNA binding motif containing 1; zinc finger CCHC-type and RNA-binding motif-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
362990, 67197, 85437 | |
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q499V6, Q8TBF4, Q9CZ96 | |
Zcrb1 | |
A 17 amino acid peptide near the carboxy terminus of human ZCRB1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction