Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZCRB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541924
Description
Peptide sequence: MSGGLAPSKS TVYVSNLPFS LTNNDLYRIF SKYGKVVKVT IMKDKDTRKS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 91%.
ZCRB1 is a nuclear protein first identified in a differential display screen involving morphine-dependence-related genes. Its expression is increased following morphine treatment and has been observed to be elevated in HepG2 cells. ZCRB1 contains a CCHC-type zing finger RNA-binding motif and can interact with the DBA-binding domain of the stem cell regulator C/EBP transcription factors. ZCRB1 was identified as one of the protein components of U11/U12 snRNPs, which are components of U12-type spliceosome and function as a molecular bridge connecting both ends of the intron, suggesting ZCRB1 may play a key role in U12-type splicing.
Specifications
| ZCRB1 | |
| Polyclonal | |
| Unconjugated | |
| Zcrb1 | |
| 2700088M22Rik; MADP1; Madp-1; MADP-1 protein; MADP-1 protein-like; MGC26805; RBM36; RGD1309851; SNRNP31; U11/U12 small nuclear ribonucleoprotein 31 kDa protein; U11/U12 snRNP 31 kDa protein; U11/U12 snRNP 31K; U11/U12-31K; ZCCHC19; Zcrb1; zinc finger CCHC-type and RNA binding motif 1; zinc finger CCHC-type and RNA binding motif containing 1; zinc finger CCHC-type and RNA-binding motif-containing protein 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 85437 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TBF4 | |
| Zcrb1 | |
| Synthetic peptide directed towards the N-terminal of human ZCRB1 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction