Learn More
ZCRB1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304697AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine
ZCRB1 is a nuclear protein first identified in a differential display screen involving morphine-dependence-related genes. Its expression is increased following morphine treatment and has been observed to be elevated in HepG2 cells. ZCRB1 contains a CCHC-type zing finger RNA-binding motif and can interact with the DBA-binding domain of the stem cell regulator C/EBP transcription factors. ZCRB1 was identified as one of the protein components of U11/U12 snRNPs, which are components of U12-type spliceosome and function as a molecular bridge connecting both ends of the intron, suggesting ZCRB1 may play a key role in U12-type splicing.Specifications
ZCRB1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8TBF4, Q9CZ96 | |
Zcrb1 | |
Between 155 and 205 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ZCRB1 | |
MADP-1, MADP1, RBM36, ZCCHC19; U11/U12 small nuclear ribonucleoprotein 31 kDa protein; U11/U12 snRNP 31 kDa protein; U11/U12 snRNP 31K; U11/U12-31K; zinc finger CCHC-type and RNA-binding motif-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
67197, 85437 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.