Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZCWPW1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596731
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596731 Supplier Invitrogen™ Supplier No. PA596731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VMKKRRNDCS QKLGVALMMA QEAEQISIQE RVNLFGFWSR FNGSNSNGER KDLQLSGLNS PGSCLEKKEK EEELEKEEGE KTDPILPIRK RVKIQTQKTK PRGLGGDAGT ADGRGRTLQR KIMKRSLGRK STAPPAPRMG RKEGQGNSDS DQPGPKKKFK APQSKALAAS FSEGKEVRTV PKNLGLSACK GACPSSAKEE PRHREPLTQE AGSVPLEDEA SSDLDLEQLM EDVGRELGQS GELQHSNSDG EDFPVALFGK; Positive Samples: SW480, BT-474, HepG2, Mouse brain.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCWPW1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZCWPW1
Gene Accession No. Q6IR42, Q9H0M4
Gene Alias Gm1053; ZCW1; Zcwpw1; zinc finger CW type with PWWP domain 1; zinc finger CW-type and PWWP domain 1; zinc finger CW-type and PWWP domain containing 1; zinc finger CW-type PWWP domain protein 1; zinc finger, CW type with PWWP domain 1; zinc finger, CW-type with PWWP domain 1
Gene Symbols ZCWPW1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 389-648 of human ZCWPW1 (NP_0604543).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381678, 55063
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.