Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZCWPW2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565247 Supplier Invitrogen™ Supplier No. PA565247
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLQNWPSWPGI LCPDRFKGKY VTYDPDGNVE EYHIEFLGDP HSRSWIKATF VGHYSITLKP EKCKNKKKWY KSALQEACLL Highest antigen sequence identity to the following orthologs - mouse 27%, rat 72%.

ZCWPW2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCWPW2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZCWPW2
Gene Accession No. Q504Y3
Gene Alias ZCW2; ZCWPW2; zinc finger CW-type and PWWP domain 2; zinc finger CW-type and PWWP domain containing 2; zinc finger CW-type PWWP domain protein 2; zinc finger, CW type with PWWP domain 2
Gene Symbols ZCWPW2
Host Species Rabbit
Immunogen Recombinant Human ZCWPW2. Recombinant protein control fragment (Product #RP-105969).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152098
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.