Learn More
Description
Western Blotting
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Gene Accession No. | BC011886 |
| Molecular Weight (g/mol) | 15.18 |
| pH Range | 6.8 |
| Plasmid | pCMV-ZD52F10 full-length |
| Quality Control Testing | Transient overexpression cell Lysate was tested with Anti-ZD52F10 antibody by western blotting |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer: 50mM Tris HCl, 2% SDS, 300mM 2-mercaptoethanol, 0.01% Bromophenol Blue, 10% Glycerol |
| Host Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.