Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDBF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565608
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565608 Supplier Invitrogen™ Supplier No. PA565608
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KISSDCDDILH LVTNQSQMIV KEISLQNARH ISLVDQSYES SSSETNFDCD ASPQSTSDYP QQSVTEVNLP KEVHIGLVDK N Highest antigen sequence identity to the following orthologs - mouse 38%, rat 43%.

This gene encodes a protein containing DBF4-type zinc finger domains. This gene is imprinted and paternally expressed in lymphocytes but is more stochastically expressed in the placenta. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDBF2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZDBF2
Gene Accession No. Q9HCK1
Gene Alias DBF4-type zinc finger-containing protein 2; KIAA1571; ZDBF2; zinc finger DBF-type containing 2; zinc finger, DBF-type containing 2
Gene Symbols ZDBF2
Host Species Rabbit
Immunogen Recombinant Human ZDBF2. Recombinant protein control fragment (Product #RP-106273).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57683
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.