Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZDHHC-12 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6958R

Description
ZDHHC12 gene ontology annotations related to this gene include peptidyl-L-cysteine S-palmitoylation; protein palmitoylation; protein targeting to membrane.Specifications
ZDHHC-12 | |
Polyclonal | |
Unconjugated | |
ZDHHC12 | |
1190004A01Rik; DHHC-12; membrane-associated DHHC12 zinc finger protein; Probable palmitoyltransferase ZDHHC12; PSEC0008; ZDHHC12; Zinc finger DHHC domain-containing protein 12; zinc finger DHHC-type containing 12; Zinc finger protein 400; zinc finger, DHHC domain containing 12; zinc finger, DHHC-type containing 12; ZNF400 | |
Rabbit | |
Protein A | |
RUO | |
84885 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96GR4 | |
ZDHHC12 | |
KLH conjugated synthetic peptide derived from human ZDHHC-12. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction