Learn More
Invitrogen™ ZDHHC1 Polyclonal Antibody

Description
Immunogen sequence: DPADANVRDK SYAGPLPIFN RSQHAHVIED LHCNLCNVDV SARSKHCS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Specifications
Specifications
| Antigen | ZDHHC1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZDHHC1 |
| Gene Accession No. | Q8WTX9 |
| Gene Alias | 4432412D04Rik; C16orf1; DHHC domain-containing cysteine-rich protein 1; DHHC-1; DHHC-domain-containing cysteine-rich protein; HSU90653; probable palmitoyltransferase ZDHHC1; ZDHHC1; Zinc finger DHHC domain-containing protein 1; zinc finger DHHC-type containing 1; zinc finger protein 377; zinc finger, DHHC domain containing 1; zinc finger, DHHC-type containing 1; ZNF377 |
| Gene Symbols | ZDHHC1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.