Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113194
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113194 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113194 Supplier Invitrogen™ Supplier No. PA5113194
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZDHHC1 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

ZDHHC1 (Zinc Finger DHHC-Type Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include protein-cysteine S-palmitoyltransferase activity and palmitoyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.4
Gene ZDHHC1
Gene Accession No. Q8R0N9, Q8WTX9
Gene Alias 4432412D04Rik; C16orf1; DHHC domain-containing cysteine-rich protein 1; DHHC-1; DHHC-domain-containing cysteine-rich protein; HSU90653; probable palmitoyltransferase ZDHHC1; ZDHHC1; Zinc finger DHHC domain-containing protein 1; zinc finger DHHC-type containing 1; zinc finger protein 377; zinc finger, DHHC domain containing 1; zinc finger, DHHC-type containing 1; ZNF377
Gene Symbols ZDHHC1
Host Species Rabbit
Immunogen Synthesized peptide derived from human ZDHC1 (81-131).
Purification Method Antigen affinity chromatography, Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29800, 70796
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.