Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZDHHC11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25659725UL
Description
ZDHHC11 Polyclonal specifically detects ZDHHC11 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ZDHHC11 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
DHHC-11, EC 2.3.1.-, FLJ13153, Zinc finger DHHC domain-containing protein 11, Zinc finger protein 399, zinc finger, DHHC domain containing 11, zinc finger, DHHC-type containing 11, ZNF399probable palmitoyltransferase ZDHHC11 | |
Rabbit | |
Affinity Purified | |
RUO | |
79844 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
ZDHHC11 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RVLRTDPRYEDVKNMNTWLLFL | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction