Learn More
Invitrogen™ ZDHHC11 Polyclonal Antibody

Description
Recommended positive controls: HCT116, MCF-7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ZDHHC11 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | ZDHHC11 |
| Gene Accession No. | Q9H8X9 |
| Gene Alias | 4933421L13Rik; DHHC-11; DHHC-containing protein 10; LOC100057809; membrane-associated DHHC11 zinc finger protein; probable palmitoyltransferase ZDHHC11; Zdhhc11; Zdhhc20; Zinc finger DHHC domain-containing protein 11; zinc finger DHHC-type containing 11; zinc finger protein 399; zinc finger, DHHC domain containing 11; zinc finger, DHHC domain containing 20; zinc finger, DHHC-type containing 11; Znf399 |
| Gene Symbols | ZDHHC11 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.