Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZDHHC11B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563500
Description
Immunogen sequence: MDTRSGSQCS VTPEAIRNNE ELVLPPRISR VNGWSLP Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 51%.
ZDHHC11B (Zinc Finger DHHC-Type Containing 11B) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includeprotein-cysteine S-palmitoyltransferase activity. An important paralog of this gene isZDHHC11.
Specifications
ZDHHC11B | |
Polyclonal | |
Unconjugated | |
ZDHHC11B | |
DHHC-11B; probable palmitoyltransferase ZDHHC11B; ZDHHC11B; Zinc finger DHHC domain-containing protein 11B; zinc finger DHHC-type containing 11B; zinc finger, DHHC-type containing 11B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
653082 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P0C7U3 | |
ZDHHC11B | |
Recombinant protein corresponding to Human ZDHHC11B. Recombinant protein control fragment (Product #RP-91241). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction