Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563705
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563705 Supplier Invitrogen™ Supplier No. PA563705
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDPGYVNVQP QPQEELKEEQ TAMVPPAIPL RRCRYCLVLQ PLRARHCREC RRCVRRY Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 93%.

ZDHHC12 gene ontology annotations related to this gene include peptidyl-L-cysteine S-palmitoylation; protein palmitoylation; protein targeting to membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZDHHC12
Gene Accession No. Q96GR4
Gene Alias 1190004A01Rik; DHHC-12; membrane-associated DHHC12 zinc finger protein; probable palmitoyltransferase ZDHHC12; PSEC0008; ZDHHC12; Zinc finger DHHC domain-containing protein 12; zinc finger DHHC-type containing 12; zinc finger DHHC-type palmitoyltransferase 12; zinc finger protein 400; zinc finger, DHHC domain containing 12; zinc finger, DHHC-type containing 12; ZNF400
Gene Symbols ZDHHC12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZDHHC12. Recombinant protein control fragment (Product #RP-101932).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84885
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.