Learn More
Invitrogen™ ZDHHC13 Monoclonal Antibody (2A4)

Description
Peptide Sequence: NGQTPLMLSA HKVIGPEPTG FLLKFNPSLN VVDKIHQNTP LHWAVAAGNV NAVDKLLEAG SSLDIQNVKG ETPLDMALQN KNQLIIHMLK TEAKMRANQK.
Specifications
Specifications
| Antigen | ZDHHC13 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2A4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZDHHC13 |
| Gene Accession No. | Q8IUH4 |
| Gene Alias | 2410004E01Rik; C530010M18; DHHC-13; fi22e09; FLJ10852; FLJ10941; Hip14l; HIP14-like; HIP14-related protein; HIP3RP; huntingtin interacting protein HIP3RP; huntingtin-interacting protein 14-related protein; Huntingtin-interacting protein HIP3RP; hypothetical protein LOC494107; kojak; MGC64994; Palmitoyltransferase ZDHHC13; probable palmitoyltransferase ZDHHC13; putative MAPK-activating protein PM03; Putative NF-kappa-B-activating protein 209; skc4; wu:fb06e01; wu:fc39g10; wu:fi22e09; Zdhhc13; zgc:101690; Zinc finger DHHC domain-containing protein 13; zinc finger DHHC-type containing 13; zinc finger DHHC-type palmitoyltransferase 13; zinc finger, DHHC domain containing 13; zinc finger, DHHC-type containing 13 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.