Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542565
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542565 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542565 Supplier Invitrogen™ Supplier No. PA542565
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MVILLLQHGA DPTLIDGEGF SSIHLAVLFQ HMPIIAYLIS KGQSVNMTDV Sequence homology: Cow: 91%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Yeast: 83%; Zebrafish: 91%.

Palmitoyltransferase for HTT and GAD2. May play a role in Mg(2+) transport. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC13
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZDHHC13
Gene Accession No. Q8IUH4
Gene Alias 2410004E01Rik; C530010M18; DHHC-13; fi22e09; FLJ10852; FLJ10941; Hip14l; HIP14-like; HIP14-related protein; HIP3RP; huntingtin interacting protein HIP3RP; huntingtin-interacting protein 14-related protein; Huntingtin-interacting protein HIP3RP; hypothetical protein LOC494107; kojak; MGC64994; Palmitoyltransferase ZDHHC13; probable palmitoyltransferase ZDHHC13; putative MAPK-activating protein PM03; Putative NF-kappa-B-activating protein 209; skc4; wu:fb06e01; wu:fc39g10; wu:fi22e09; Zdhhc13; zgc:101690; Zinc finger DHHC domain-containing protein 13; zinc finger DHHC-type containing 13; zinc finger DHHC-type palmitoyltransferase 13; zinc finger, DHHC domain containing 13; zinc finger, DHHC-type containing 13
Gene Symbols ZDHHC13
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZDHHC13 (aa 131-180).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54503
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.