Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582635
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582635 Supplier Invitrogen™ Supplier No. PA582635
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSHERISLQK QSKHMKQTLS LRKTPYNLGF MQNLADFFQC GCFGLVKPCV VDWTSQYTMV FHPARE Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 92%.

Palmitoyltransferase for HTT and GAD2. May play a role in Mg(2+) transport. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC13
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZDHHC13
Gene Accession No. Q8IUH4
Gene Alias 2410004E01Rik; C530010M18; DHHC-13; fi22e09; FLJ10852; FLJ10941; Hip14l; HIP14-like; HIP14-related protein; HIP3RP; huntingtin interacting protein HIP3RP; huntingtin-interacting protein 14-related protein; Huntingtin-interacting protein HIP3RP; hypothetical protein LOC494107; kojak; MGC64994; Palmitoyltransferase ZDHHC13; probable palmitoyltransferase ZDHHC13; putative MAPK-activating protein PM03; Putative NF-kappa-B-activating protein 209; skc4; wu:fb06e01; wu:fc39g10; wu:fi22e09; Zdhhc13; zgc:101690; Zinc finger DHHC domain-containing protein 13; zinc finger DHHC-type containing 13; zinc finger DHHC-type palmitoyltransferase 13; zinc finger, DHHC domain containing 13; zinc finger, DHHC-type containing 13
Gene Symbols ZDHHC13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZDHHC13. Recombinant protein control fragment (Product #RP-91299).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54503
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.