Learn More
ZDHHC14 Polyclonal Antibody

Description
ZDHHC14 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | ZDHHC14 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ZDHHC14 |
| Gene Accession No. | Q8BQQ1, Q8IZN3 |
| Gene Alias | B530001K09Rik; Dhhc14; DHHC-14; membrane-associated DHHC14 zinc finger protein; NEW1 domain containing protein; NEW1 domain-containing protein; NEW1CP; probable palmitoyltransferase ZDHHC14; probable palmitoyltransferase ZDHHC14; LOW QUALITY PROTEIN: probable palmitoyltransferase ZDHHC14; si:ch211-199l3.1; ZDHHC14; Zinc finger DHHC domain-containing protein 14; zinc finger DHHC-type containing 14; zinc finger DHHC-type palmitoyltransferase 14; zinc finger, DHHC domain containing 14; zinc finger, DHHC-type containing 14 |
| Gene Symbols | ZDHHC14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.