Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZDHHC14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569184
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
ZDHHC14 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane; protein-cysteine S-palmitoyltransferase activity.
Specifications
ZDHHC14 | |
Polyclonal | |
Unconjugated | |
ZDHHC14 | |
B530001K09Rik; Dhhc14; DHHC-14; membrane-associated DHHC14 zinc finger protein; NEW1 domain containing protein; NEW1 domain-containing protein; NEW1CP; probable palmitoyltransferase ZDHHC14; probable palmitoyltransferase ZDHHC14; LOW QUALITY PROTEIN: probable palmitoyltransferase ZDHHC14; si:ch211-199l3.1; ZDHHC14; Zinc finger DHHC domain-containing protein 14; zinc finger DHHC-type containing 14; zinc finger DHHC-type palmitoyltransferase 14; zinc finger, DHHC domain containing 14; zinc finger, DHHC-type containing 14 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79683 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IZN3 | |
ZDHHC14 | |
synthetic peptide directed towards the N terminal region of human ZDHHC14. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction