Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598601
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598601 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598601 Supplier Invitrogen™ Supplier No. PA598601
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZDHHC14 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

ZDHHC14 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane; protein-cysteine S-palmitoyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC14
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZDHHC14
Gene Accession No. Q8IZN3
Gene Alias B530001K09Rik; Dhhc14; DHHC-14; membrane-associated DHHC14 zinc finger protein; NEW1 domain containing protein; NEW1 domain-containing protein; NEW1CP; probable palmitoyltransferase ZDHHC14; probable palmitoyltransferase ZDHHC14; LOW QUALITY PROTEIN: probable palmitoyltransferase ZDHHC14; si:ch211-199l3.1; ZDHHC14; Zinc finger DHHC domain-containing protein 14; zinc finger DHHC-type containing 14; zinc finger DHHC-type palmitoyltransferase 14; zinc finger, DHHC domain containing 14; zinc finger, DHHC-type containing 14
Gene Symbols ZDHHC14
Host Species Rabbit
Immunogen Recombinant Human Probable palmitoyltransferase ZDHHC14 protein (328-453AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79683
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.