Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589841
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589841 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589841 Supplier Invitrogen™ Supplier No. PA589841
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSRNKTTLEA FCTPVFTSGP EKNGFNLGFI KNIQQVFGDK KKFWLIPIGS SPGDGHSFPM RSMNESQNPL LANEETWEDN EDDNQDYPEG SSSLAVETET; Positive Samples: U-87MG, HepG2; Cellular Location: Membrane, Multi-pass membrane protein.

The protein encoded by this gene belongs to the DHHC palmitoyltransferase family. Mutations in this gene are associated with mental retardatio X-linked type 91 (MRX91). Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC15
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZDHHC15
Gene Accession No. Q2TGJ4, Q96MV8
Gene Alias 6030457O13Rik; DHHC-15; membrane-associated DHHC15 zinc finger protein; MRX91; Palmitoyltransferase ZDHHC15; UNQ1969/PRO4501; Zdhhc15; Zinc finger DHHC domain-containing protein 15; zinc finger DHHC-type containing 15; zinc finger, DHHC domain containing 15; zinc finger, DHHC-type containing 15
Gene Symbols ZDHHC15
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 238-337 of human ZDHHC15 (NP_6594061).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158866, 317235
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.