Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZDHHC16 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP189041
Description
ZDHHC16 Polyclonal specifically detects ZDHHC16 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZDHHC16 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
DHHC domain containing 16, EC 2.3.1, EC 2.3.1.-, zinc finger, DHHC-type containing 16 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ZDHHC16 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VLISRGETSIERHINKKERRRLQAKGRVFRNPYNYGCLDNWKVFLGVDTGRHWLTRVLLPSSHLPHGNGMSWEPPPWVTA | |
0.1 mL | |
Apoptosis | |
84287 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction