Learn More
Invitrogen™ ZDHHC17 Polyclonal Antibody

Description
Immunogen sequence: RKEGFNTKMA DGPDEYDTEA GCVPLLHPEE IKPQSHYNHG YGEPLGRKTH IDDYSTWDIV KATQYGIYER CRELVEAGYD VR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 88%.
Specifications
Specifications
| Antigen | ZDHHC17 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZDHHC17 |
| Gene Accession No. | Q8IUH5 |
| Gene Alias | A230053P19Rik; BB187739; D130071N24Rik; DHHC-17; Hip14; HIP-14; HIP3; HIP-3; HSPC294; huntingtin interacting protein 14; huntingtin interacting protein 3; Huntingtin interacting protein H; huntingtin yeast partner H; Huntingtin-interacting protein 14; huntingtin-interacting protein 3; Huntingtin-interacting protein H; HYPH; Kiaa0946; membrane-associated DHHC17 zinc finger protein; palmito; palmitoyltransferase ZDHHC17; Putative MAPK-activating protein PM11; putative NF-kappa-B-activating protein 205; ZDHHC17; Zinc finger DHHC domain-containing protein 17; zinc finger DHHC-type containing 17; zinc finger, DHHC domain containing 17; zinc finger, DHHC-type containing 17 |
| Gene Symbols | ZDHHC17 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.