Learn More
Invitrogen™ ZDHHC17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593227
Description
Immunogen sequence: TSIVAYLIAK GQDVDMMDQN GMTPLMWAAY RTHSVDPTRL LLTFNVSVNL GDKYHKNTAL HWAVLAGNTT VISLLLEAGA NVDAQNIKGE SALDLAKQRK NVWMINHLQE ARQAKGYDNP SFLRKLKADK EFRQKVMLGT P; Positive Samples: HL60, SKOV3, 293T; Cellular Location: Cytoplasmic vesicle membrane, Golgi apparatus membrane, Multi-pass membrane protein.
Palmitoyltransferase specific for a subset of neuronal proteins, including SNAP25, DLG4/PSD95, GAD2, SYT1 and HD. May be involved in the sorting or targeting of critical proteins involved in the initiating events of endocytosis at the plasma membrane. Has transforming activity. Mediates Mg(2+) transport.
Specifications
ZDHHC17 | |
Polyclonal | |
Unconjugated | |
ZDHHC17 | |
A230053P19Rik; BB187739; D130071N24Rik; DHHC-17; Hip14; HIP-14; HIP3; HIP-3; HSPC294; huntingtin interacting protein 14; huntingtin interacting protein 3; Huntingtin interacting protein H; huntingtin yeast partner H; Huntingtin-interacting protein 14; huntingtin-interacting protein 3; Huntingtin-interacting protein H; HYPH; Kiaa0946; membrane-associated DHHC17 zinc finger protein; palmito; palmitoyltransferase ZDHHC17; Putative MAPK-activating protein PM11; putative NF-kappa-B-activating protein 205; ZDHHC17; Zinc finger DHHC domain-containing protein 17; zinc finger DHHC-type containing 17; zinc finger, DHHC domain containing 17; zinc finger, DHHC-type containing 17 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23390, 320150, 366889 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
E9PTT0, Q80TN5, Q8IUH5 | |
ZDHHC17 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 170-310 of human ZDHHC17 (NP_0561512). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.