Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518336
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518336 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518336 Supplier Invitrogen™ Supplier No. PA518336
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

Palmitoyltransferase specific for a subset of neuronal proteins, including SNAP25, DLG4/PSD95, GAD2, SYT1 and HD. May be involved in the sorting or targeting of critical proteins involved in the initiating events of endocytosis at the plasma membrane. Has transforming activity. Mediates Mg(2+) transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ZDHHC17
Gene Accession No. Q80TN5, Q8IUH5
Gene Alias A230053P19Rik; BB187739; D130071N24Rik; DHHC-17; Hip14; HIP-14; HIP3; HIP-3; HSPC294; huntingtin interacting protein 14; huntingtin interacting protein 3; Huntingtin interacting protein H; huntingtin yeast partner H; Huntingtin-interacting protein 14; huntingtin-interacting protein 3; Huntingtin-interacting protein H; HYPH; Kiaa0946; membrane-associated DHHC17 zinc finger protein; palmito; palmitoyltransferase ZDHHC17; Putative MAPK-activating protein PM11; putative NF-kappa-B-activating protein 205; ZDHHC17; Zinc finger DHHC domain-containing protein 17; zinc finger DHHC-type containing 17; zinc finger, DHHC domain containing 17; zinc finger, DHHC-type containing 17
Gene Symbols ZDHHC17
Host Species Goat
Immunogen Synthetic peptide sequence (CYDQISGSGYQLV) corresponding to the C-terminus amino acids of ZDHHC17 (aa 621-632).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23390, 320150
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.