Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC17 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7386 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7386 Supplier Bioss Supplier No. BS6628R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Palmitoyltransferase specific for a subset of neuronal proteins, including SNAP25, DLG4/PSD95, GAD2, SYT1 and HD. May be involved in the sorting or targeting of critical proteins involved in the initiating events of endocytosis at the plasma membrane. Has transforming activity. Mediates Mg(2+) transport.

Specifications

Antigen ZDHHC17
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZDHHC17
Gene Accession No. Q8IUH5
Gene Alias A230053P19Rik; BB187739; D130071N24Rik; DHHC-17; Hip14; HIP-14; HIP3; HIP-3; HSPC294; huntingtin interacting protein 14; huntingtin interacting protein 3; Huntingtin interacting protein H; Huntingtin yeast partner H; Huntingtin-interacting protein 14; Huntingtin-interacting protein 3; Huntingtin-interacting protein H; HYPH; Kiaa0946; membrane-associated DHHC17 zinc finger protein; palmito; Palmitoyltransferase ZDHHC17; Putative MAPK-activating protein PM11; Putative NF-kappa-B-activating protein 205; Zdhhc17; Zinc finger DHHC domain-containing protein 17; zinc finger DHHC-type containing 17; zinc finger, DHHC domain containing 17; zinc finger, DHHC-type containing 17
Gene Symbols ZDHHC17
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZDHHC17.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23390
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.