Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103997
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103997 Supplier Invitrogen™ Supplier No. PA5103997
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZDHHC18.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. ZDHHC1 (zinc finger, DHHC-type containing 1), also known as ZNF377 (zinc finger protein 377) or C16orf1, is a 485 amino acid multi-pass membrane protein that contains one DHHC-type zinc finger. Expressed in fetal heart, lung and kidney and also present in adult lung and pancreas, ZDHHC1 functions as a palmitoyltransferase that catalyzes the conversion of palmitoyl-CoA and protein-cysteine to S-palmitoyl protein and CoA. Like ZDHHC1, ZDHHC5, ZDHHC6, ZDHHC7 and ZDHHC18 each contain one DHHC-type zinc finger through which they convey palmitoyltransferase activity against a broad range of substrates, including H-Ras, SNAP 25 and GABAA R proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC18
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZDHHC18
Gene Accession No. Q2TGJ1, Q5Y5T2, Q9NUE0
Gene Alias DHHC18; DHHC-18; membrane-associated DHHC18 zinc finger protein; palmitoyltransferase ZDHHC18; Zdhhc18; Zinc finger DHHC domain-containing protein 18; zinc finger DHHC-type containing 18; zinc finger, DHHC domain containing 18; zinc finger, DHHC-type containing 18
Gene Symbols ZDHHC18
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZDHHC18(Accession Q9NUE0), corresponding to amino acid residues A42-G92.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362613, 503610, 84243
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.