Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568526
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568526 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568526 Supplier Invitrogen™ Supplier No. PA568526
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Horse: 92%; Human: 100%; Mouse: 91%; Pig: 86%; Rat: 91%.

ZDHHC19 gene ontology annotations related to this gene include peptidyl-L-cysteine S-palmitoylation; protein targeting to membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC19
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZDHHC19
Gene Accession No. Q8WVZ1
Gene Alias DHHC19; DHHC-19; Gm1744; Gm616; LOC100069974; LOW QUALITY PROTEIN: probable palmitoyltransferase ZDHHC19; membrane-associated DHHC19 zinc finger protein; palmitoyltransferase ZDHHC19; probable palmitoyltransferase ZDHHC19; RGD1560310; Unknown (protein for MGC:137377); Zdhhc19; Zinc finger DHHC domain-containing protein 19; zinc finger DHHC-type containing 19; zinc finger DHHC-type palmitoyltransferase 19; zinc finger, DHHC domain containing 19; zinc finger, DHHC-type containing 19
Gene Symbols ZDHHC19
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human ZDHHC19.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131540
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.